DL-Panthenol 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Panthenol, DL-Pantothenol, 2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide

B25114-18 B25114-30
CAAAAB25114-0BEA 669.13 CAD
CAAAAB25114-0B CAAAAB25114-18 CAAAAB25114-30
DL-Panthenol 99%
DL-Panthenol

Purity: 99% CAS No.: 16485-10-2 Molecular Formula: C9H19NO4 Formula Weight: 205.26 Storage Temperature: 2°C to 8°C Melting Point: 66-69° MDL No.: MFCD00002944

Formula: C₉H₁₉NO₄
MW: 205.25 g/mol
Melting Pt: 66…69 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002944
CAS Number: 16485-10-2
EINECS: 240-540-6
Merck Index: 14,2947

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR