Nalidixic acid 99%

Supplier: Thermo Scientific Chemicals
Danger

B25096-06 B25096-14 B25096-22
CAAAAB25096-06EA 49.3 CAD
CAAAAB25096-06 CAAAAB25096-14 CAAAAB25096-22
Nalidixic acid 99%
Nalidixic acid

Purity: 99% CAS No.: 389-08-2 Molecular Formula: C12H12N2O3 Formula Weight: 232.24 Melting Point: 228-230° Beilstein Reference: 750515 Merck Reference: 14,6359 MDL No.: MFCD00006884

Formula: C₁₂H₁₂N₂O₃
MW: 232.24 g/mol
Melting Pt: 228…230 °C
Storage Temperature: Ambient
MDL Number: MFCD00006884
CAS Number: 389-08-2
EINECS: 206-864-7
Merck Index: 14,6359

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR