Creatine (N-amidinosarcosine) monohydrate 99%
Supplier: Thermo Scientific Chemicals
B25009-0E
B25009-22
B25009-36
CAAAAB25009-0EEA
820.02
CAD
CAAAAB25009-0E
CAAAAB25009-22
CAAAAB25009-36
Creatine (N-amidinosarcosine) monohydrate 99%
Creatine (N-Amidinosarcosine) monohydrate
Purity: 99%
CAS No.: 6020-87-7
Molecular Formula: C4H9N3O2.H2O
Formula Weight: 149.15 (131.13anhy)
Odor: Weak
pH: 6.9
Melting Point: ca 292° dec.
Beilstein Reference: 907175
Merck Reference: 14,2568
MDL No.: MFCD00071582
Formula:
C₄H₉N₃O₂·1H₂O Melting Pt: ∼ 292 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00071582 CAS Number: 6020-87-7 EINECS: 200-306-6 Merck Index: 14,2568 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...