Methyl propionylacetate 99%
Supplier: Thermo Scientific Chemicals
Warning
|
B24848-14
B24848-22
B24848-36
CAAAAB24848-14EA
101.59
CAD
CAAAAB24848-14
CAAAAB24848-22
CAAAAB24848-36
Methyl propionylacetate 99%
Methyl propionylacetate
Purity: 99%
CAS No.: 30414-53-0
Molecular Formula: C6H10O3
Formula Weight: 130.14
Flash Point: 71°(159°F)
Refractive Index: 1.4220
Boiling Point: 73-74°/5mm
Beilstein Reference: 1753684
MDL No.: MFCD00011705
Formula:
C₆H₁₀O₃ Boiling Pt: 73…74 °C/5 mm Density: 1,037 Flash Pt: 71 °C(159 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00011705 CAS Number: 30414-53-0 EINECS: 250-184-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...