Didecyldimethylammonium bromide ≥80%, aqueous gel
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24832-22
B24832-36
CAAAAB24832-22EA
101.44
CAD
CAAAAB24832-22
CAAAAB24832-36
Didecyldimethylammonium bromide ≥80%, aqueous gel
Didecyldimethylammonium bromide
CAS No.: 2390-68-3
Molecular Formula: C22H48BrN
Formula Weight: 406.54
Refractive Index: 1.4570
MDL No.: MFCD00012194
Formula:
C₂₂H₄₈BrN Storage Temperature: Ambient |
MDL Number:
MFCD00012194 CAS Number: 2390-68-3 EINECS: 219-234-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...