(+)-Dibenzoyl-D-tartaric acid, anhydrous 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2S,3S)-(+)-Di-O-benzoyltartaric acid, (2S,3S)-2,3-Bis(benzoyloxy)succinic acid

B24754-09 B24754-18 B24754-30
CAAAAB24754-09EA 44.87 CAD
CAAAAB24754-09 CAAAAB24754-18 CAAAAB24754-30
(+)-Dibenzoyl-D-tartaric acid, anhydrous 99%
(+)-Dibenzoyl-D-tartaric acid

For chiral resolution.


Purity: 99% CAS No.: 17026-42-5 Molecular Formula: C18H14O8 Formula Weight: 358.31 Melting Point: 154-156° Beilstein Reference: 2227343

Formula: C₁₈H₁₄O₈
MW: 358.31 g/mol
Melting Pt: 154…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00063222
CAS Number: 17026-42-5
EINECS: 241-097-1

Order Now

Specification Test Results

Optical rotation +120° (c=5 in methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR