4-(Dimethylamino)cinnamaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24741-06
B24741-14
CAAAAB24741-06EA
134.93
CAD
CAAAAB24741-06
CAAAAB24741-14
4-(Dimethylamino)cinnamaldehyde 98%
4-(Dimethylamino)cinnamaldehyde
Purity: 98%
CAS No.: 6203-18-5
Molecular Formula: C11H13NO
Formula Weight: 175.23
Melting Point: 137-144°
Beilstein Reference: 972369
Formula:
C₁₁H₁₃NO Melting Pt: 137…144 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007002 CAS Number: 6203-18-5 EINECS: 228-267-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...