(R)-(+)-2-Pyrrolidinone-5-carboxylic acid 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: D-Pyr-OH, (R)-(+)-2-Pyrrolidone-5-carboxylic acid

B24678-06 B24678-14 B24678-22
CAAAAB24678-06EA 116.29 CAD
CAAAAB24678-06 CAAAAB24678-14 CAAAAB24678-22
(R)-(+)-2-Pyrrolidinone-5-carboxylic acid 98+%
(R)-(+)-2-Pyrrolidinone-5-carboxylic acid

Purity: 98+% CAS No.: 4042-36-8 Molecular Formula: C5H7NO3 Formula Weight: 129.12 Melting Point: 158-160° Beilstein Reference: 82133

Formula: C₅H₇NO₃
MW: 129.12 g/mol
Melting Pt: 158…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00066212
CAS Number: 4042-36-8
EINECS: 223-735-0

Order Now

Specification Test Results

Optical rotation +10.5° (c=5 in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR