3-Methylbenzo[b]thiophene-2-carboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24624-03
B24624-06
B24624-14
CAAAB24624-03EA
83.11
CAD
CAAAB24624-03
CAAAB24624-06
CAAAB24624-14
3-Methylbenzo[b]thiophene-2-carboxylic acid 97%
3-Methylbenzo[b]thiophene-2-carboxylic acid
Purity: 97%
CAS No.: 3133-78-6
Molecular Formula: C10H8O2S
Formula Weight: 192.24
Melting Point: 241-244°
MDL No.: MFCD01830305
Formula:
C₁₀H₈O₂S Melting Pt: 241…244 °C Storage Temperature: Ambient |
MDL Number:
MFCD01830305 CAS Number: 3133-78-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...