2-Bromo-3,3,3-trifluoropropene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Bromo-3,3,3-trifluoro-1-propene
B24614-06
B24614-14
CAAAB24614-06EA
125.77
CAD
CAAAB24614-06
CAAAB24614-14
2-Bromo-3,3,3-trifluoropropene 97%
2-Bromo-3,3,3-trifluoropropene
Purity: 97%
CAS No.: 1514-82-5
Molecular Formula: C3H2BrF3
Formula Weight: 174.95
Refractive Index: 1.3530
Formula:
C₃H₂BrF₃ Storage Temperature: Ambient |
MDL Number:
MFCD00077469 CAS Number: 1514-82-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...