2-Methyl-2-oxazoline 99%

Supplier: Thermo Scientific Chemicals
Danger

B24612-14 B24612-22
CAAAB24612-14EA 58.14 CAD
CAAAB24612-14 CAAAB24612-22
2-Methyl-2-oxazoline 99%
2-Methyl-2-oxazoline

Purity: 99% CAS No.: 1120-64-5 Molecular Formula: C4H7NO Formula Weight: 85.11 Flash Point: 20°(68°F) Refractive Index: 1.4340 Boiling Point: 109-111° Beilstein Reference: 104227 UN No.: 1993 MDL No.: MFCD00005298

Formula: C₄H₇NO
Boiling Pt: 109…111 °C
Density: 1.01
Flash Pt: 20 °C(68 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005298
CAS Number: 1120-64-5
EINECS: 214-316-3
UN: 1993
ADR: "3",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR