N,N-Dimethylthioformamide 97%

Supplier: Thermo Scientific Chemicals
Warning

B24529-06 B24529-14 B24529-22
CAAAB24529-06EA 80.9 CAD
CAAAB24529-06 CAAAB24529-14 CAAAB24529-22
N,N-Dimethylthioformamide 97%
N,N-Dimethylthioformamide

Purity: 97% CAS No.: 758-16-7 Molecular Formula: C3H7NS Formula Weight: 89.15 Boiling Point: 58-60°/1mm MDL No.: MFCD00004943

Formula: C₃H₇NS
Boiling Pt: 58…60 °C/1 mm
Storage Temperature: Ambient
MDL Number: MFCD00004943
CAS Number: 758-16-7
EINECS: 212-060-7
UN: 2810
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR