(±)-2-Chloropropionyl chloride 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B24503-14
B24503-22
B24503-36
CAAAB24503-14EA
76.31
CAD
CAAAB24503-14
CAAAB24503-22
CAAAB24503-36
(±)-2-Chloropropionyl chloride 96%
(±)-2-Chloropropionyl chloride
Purity: 96%
CAS No.: 7623-09-8
Molecular Formula: CH3CHClCOCl
Formula Weight: 126.97
Flash Point: 31°(88°F)
Refractive Index: 1.4400
Boiling Point: 109-111°
UN No.: 2920
MDL No.: MFCD00000862
Formula:
C₃H₄Cl₂O Boiling Pt: 109…111 °C Density: 1.308 Flash Pt: 31 °C(88 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000862 CAS Number: 7623-09-8 EINECS: 231-540-7 UN: 2920 ADR: "8",II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...