2-Methoxypropene 95% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
Stabilization:
Stabilised with potassium carbonate ∼0.5%
B24083-18
B24083-30
CAAAB24083-18EA
74.89
CAD
CAAAB24083-18
CAAAB24083-30
2-Methoxypropene 95% stabilized
2-Methoxypropene
Fieser: 2,230 5,360 12,317
Purity: 95%
CAS No.: 116-11-0
Molecular Formula: C4H8O
Formula Weight: 72.11
Storage Temperature: 2°C to 8°C
Flash Point: -29°(-20°F)
Refractive Index: 1.3835
Odor: Ether-like
Boiling Point: 35-36°
Beilstein Reference: 1734635
UN No.: 1993
MDL No.: MFCD00014929
Formula:
C₄H₈O Boiling Pt: 35…36 °C Density: 0.758 Flash Pt: -29 °C(-20 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014929 CAS Number: 116-11-0 EINECS: 204-125-3 UN: 1993 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...