Saccharin 98+%
Supplier: Thermo Scientific Chemicals
B23938-0E
B23938-22
B23938-36
CAAAB23938-0EEA
526.14
CAD
CAAAB23938-0E
CAAAB23938-22
CAAAB23938-36
Saccharin 98+%
Saccharin
An artificial sweetener which is an inhibitor for phosphotransferase activity
Purity: 98+%
CAS No.: 81-07-2
Molecular Formula: C7H5NO3S
Formula Weight: 183.18
Melting Point: 227-230°
Beilstein Reference: 6888
Merck Reference: 14,8311
MDL No.: MFCD00005866
Formula:
C₇H₅NO₃S Melting Pt: 227…230 °C Density: 0.828 Storage Temperature: Ambient |
MDL Number:
MFCD00005866 CAS Number: 81-07-2 EINECS: 201-321-0 Merck Index: 14,8311 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...