Tetraethylene glycol 99%
Supplier: Thermo Scientific Chemicals
B23880-0F
B23880-AP
CAAAB23880-0FEA
178.54
CAD
CAAAB23880-0F
CAAAB23880-AP
Tetraethylene glycol 99%
Tetraethylene glycol
Used as a solvent to dissolve water-insoluble compounds
Purity: 99%
CAS No.: 112-60-7
Molecular Formula: O(CH2CH2OCH2CH2OH)2
Formula Weight: 194.23
Flash Point: 193°(379°F)
Refractive Index: 1.4590
Odor: Odorless
pH: 8.5
Melting Point: -4°
Boiling Point: 313-314°
Beilstein Reference: 1634320
MDL No.: MFCD00002879
Formula:
C₈H₁₈O₅ Boiling Pt: 313…314 °C Melting Pt: –4 °C Density: 1.125 Flash Pt: 193 °C(379 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002879 CAS Number: 112-60-7 EINECS: 203-989-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...