N-Methyl-2,2,2-trifluoroacetamide 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23851-06
B23851-14
CAAAB23851-06EA
55.14
CAD
CAAAB23851-06
CAAAB23851-14
N-Methyl-2,2,2-trifluoroacetamide 98+%
N-Methyl-2,2,2-trifluoroacetamide
Purity: 98+%
CAS No.: 815-06-5
Molecular Formula: C3H4F3NO
Formula Weight: 127.07
Flash Point: 73°(163°F)
Melting Point: 49-52°
Boiling Point: 156-157°
Beilstein Reference: 1703392
MDL No.: MFCD00009670
Formula:
C₃H₄F₃NO Boiling Pt: 156…157 °C Melting Pt: 49…52 °C Flash Pt: 73 °C(163 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009670 CAS Number: 815-06-5 EINECS: 212-417-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...