N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide (MTBSTFA) 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-(tert-Butyldimethylsilyl)-2,2,2-trifluoro-N-methylacetamide, MBDSTFA

B23760-03 B23760-06 B23760-14
CAAAB23760-03EA 68.41 CAD
CAAAB23760-03 CAAAB23760-06 CAAAB23760-14
N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide (MTBSTFA) 97%
N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide (MTBSTFA)

Purity: 97% CAS No.: 77377-52-7 Molecular Formula: C9H18F3NOSi Formula Weight: 241.33 Flash Point: 52°(125°F) Refractive Index: 1.4020 Boiling Point: 168-170° Beilstein Reference: 3606546 UN No.: 1993 MDL No.: MFCD00009671

Formula: C₉H₁₈F₃NOSi
MW: 241.33 g/mol
Boiling Pt: 168…170 °C
Density: 1.023
Flash Pt: 52 °C(125 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009671
CAS Number: 77377-52-7
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR