(±)-2-Bromopropionyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

B23662-09 B23662-18
CAAAB23662-09EA 114.87 CAD
CAAAB23662-09 CAAAB23662-18 CAAAB23662-30
(±)-2-Bromopropionyl chloride 98%
(±)-2-Bromopropionyl chloride

Purity: 98% CAS No.: 7148-74-5 Molecular Formula: CH3CHBrCOCl Formula Weight: 171.43 Flash Point: 51°(125°F) Refractive Index: 1.4780 Boiling Point: 131-133° Beilstein Reference: 773800 UN No.: 2920 MDL No.: MFCD00000711

Formula: C₃H₄BrClO
Boiling Pt: 131…133 °C
Density: 1.700
Flash Pt: 51 °C (125 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000711
CAS Number: 7148-74-5
EINECS: 230-466-2
UN: 2920
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR