N-Acetyl-L-glutamic acid 99%
Supplier: Thermo Scientific Chemicals
B23621-14
B23621-22
CAAAB23621-14EA
45.35
CAD
CAAAB23621-14
CAAAB23621-22
N-Acetyl-L-glutamic acid 99%
N-Acetyl-L-glutamic acid
Purity: 99%
CAS No.: 1188-37-0
Molecular Formula: C7H11NO5
Formula Weight: 189.17
Storage Temperature: 2°C to 8°C
Melting Point: 194-199°
Beilstein Reference: 1727473
Optical Rotation: -16.5° (c=1 in water)
MDL No.: MFCD00002802
Formula:
C₇H₁₁NO₅ Melting Pt: 194…199 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002802 CAS Number: 1188-37-0 EINECS: 214-708-4 |
Specification Test Results
Optical rotation | -16.5° (c=1 in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...