1-Dodecylpyridinium chloride hydrate 98%

Supplier: Thermo Scientific Chemicals
Warning

B23492-14 B23492-22
CAAAAB23492-14EA 91.64 CAD
CAAAAB23492-14 CAAAAB23492-22
1-Dodecylpyridinium chloride hydrate 98%
1-Dodecylpyridinium chloride hydrate

Purity: 98% CAS No.: 207234-02-4 Molecular Formula: C17H30ClN.xH2O Formula Weight: 283.89(anhy) Melting Point: 88-90° MDL No.: MFCD00150002

Formula: C₁₇H₃₀ClN·xH₂O
Melting Pt: 88…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00150002
CAS Number: 207234-02-4
EINECS: 203-232-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR