3-Hydroxy-2,4,6-triiodobenzoic acid 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: HTBA

B23355-06 B23355-14
CAAAB23355-06EA 120.87 CAD
CAAAB23355-06 CAAAB23355-14 CAAAB23355-22
3-Hydroxy-2,4,6-triiodobenzoic acid 97%
3-Hydroxy-2,4,6-triiodobenzoic acid

Purity: 97% CAS No.: 53279-72-4 Molecular Formula: C7H3I3O3 Formula Weight: 515.81 Melting Point: 211-215° Beilstein Reference: 2113494

Formula: C₇H₃I₃O₃
MW: 515.81 g/mol
Melting Pt: 211…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00020305
CAS Number: 53279-72-4
EINECS: 258-457-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR