Diethyl propylmalonate 99%
Supplier: Thermo Scientific Chemicals
Warning
|
B23304-14
B23304-22
CAAAB23304-14EA
90.85
CAD
CAAAB23304-14
CAAAB23304-22
Diethyl propylmalonate 99%
Diethyl propylmalonate
Purity: 99%
CAS No.: 2163-48-6
Molecular Formula: C10H18O4
Formula Weight: 202.25
Flash Point: 91°(197°F)
Refractive Index: 1.4205
Boiling Point: 221-222°
MDL No.: MFCD00009168
Formula:
C₁₀H₁₈O₄ Boiling Pt: 221…222 °C Density: 0.987 Flash Pt: 91 °C(197 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009168 CAS Number: 2163-48-6 EINECS: 218-492-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...