4-Methyl-5-nonanol 98%
Supplier: Thermo Scientific Chemicals
B23167-09
B23167-18
B23167-30
CAAAB23167-09EA
95.12
CAD
CAAAB23167-09
CAAAB23167-18
CAAAB23167-30
4-Methyl-5-nonanol 98%
4-Methyl-5-nonanol
Purity: 98%
CAS No.: 154170-44-2
Molecular Formula: C10H22O
Formula Weight: 158.29
Refractive Index: 1.4360
Formula:
C₁₀H₂₂O MW: 158.29 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00046696 CAS Number: 154170-44-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...