N,N-Diphenylcarbamoyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

B23116-14 B23116-22
CAAAB23116-14EA 95.12 CAD
CAAAB23116-14 CAAAB23116-22
N,N-Diphenylcarbamoyl chloride 98%
N,N-Diphenylcarbamoyl chloride

Purity: 98% CAS No.: 83-01-2 Molecular Formula: C13H10ClNO Formula Weight: 231.68 Odor: Irritating Melting Point: 82-86° Beilstein Reference: 515312 UN No.: 3261 MDL No.: MFCD00000633

Formula: C₁₃H₁₀ClNO
Melting Pt: 82…86 °C
Storage Temperature: Ambient
MDL Number: MFCD00000633
CAS Number: 83-01-2
EINECS: 201-450-2
UN: 3261
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR