(-)-Di-p-toluoyl-L-tartaric acid 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Di-p-toluoyl-L-tartaric acid, (-)-O,O'-Di-p-toluoyl-L-tartaric acid, (2R,3R)-(-)-Di-O-4-Toluoyl-L-Tartaric Acid

B22972-14 B22972-22
CAAAB22972-06EA 71.57 CAD
CAAAB22972-06 CAAAB22972-14 CAAAB22972-22
(-)-Di-p-toluoyl-L-tartaric acid 98%
(-)-Di-p-toluoyl-L-tartaric acid

Purity: 98% CAS No.: 32634-66-5 Molecular Formula: C20H18O8 Formula Weight: 386.36 Melting Point: 166-171° Beilstein Reference: 2022481

Formula: C₂₀H₁₈O₈
Melting Pt: 166…171 °C
Storage Temperature: Ambient
MDL Number: MFCD00064440
CAS Number: 32634-66-5
EINECS: 251-131-7

Order Now

Specification Test Results

Optical rotation -139° (c=1 in ethanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR