α-Acetyl-γ-butyrolactone 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
alpha-Acetyl-gamma-hydroxybutyric acid Lactone, α-Acetyl-γ-hydroxybutyric acid Lactone, 3-Acetyldihydrofuran-2(3H)-one, 2-Acetyl-γ-butyrolactone, 2-Acetylbutyrolactone, alpha-Acetyl-gamma-butyrolactone, α-Acetylbutyrolactone, 3-Acetyl-4,5-dihydro-2(3H)-furanone
Purity: 98+%
CAS No.: 517-23-7
Molecular Formula: C6H8O3
Formula Weight: 128.13
Flash Point: 135°(275°F)
Refractive Index: 1.4590
Boiling Point: 107-108°/5mm
Beilstein Reference: 112676
Merck Reference: 14,83
MDL No.: MFCD00005394
Formula:
C₆H₈O₃ Boiling Pt: 107…108 °C/5 mm Density: 1.190 Flash Pt: 135 °C(275 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005394 CAS Number: 517-23-7 EINECS: 208-235-2 Merck Index: 14,83 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...