(±)-Tetrahydro-2-furoic acid 97+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B22927-14
B22927-22
CAAAB22927-14EA
99.54
CAD
CAAAB22927-14
CAAAB22927-22
(±)-Tetrahydro-2-furoic acid 97+%
(±)-Tetrahydro-2-furoic acid
Purity: 97+%
CAS No.: 16874-33-2
Molecular Formula: C5H8O3
Formula Weight: 116.12
Flash Point: 139°(282°F)
Refractive Index: 1.4600
Odor: Pungent
Boiling Point: 244°
Beilstein Reference: 109671
UN No.: 3265
MDL No.: MFCD00022473
Formula:
C₅H₈O₃ Boiling Pt: 244 °C Density: 1.213 Flash Pt: 139 °C(282 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00022473 CAS Number: 16874-33-2 UN: 3265 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...