1-Naphthyl phosphate disodium salt monohydrate 99%

Supplier: Thermo Scientific Chemicals
Warning

B22866-03 B22866-06 B22866-14
CAAAB22866-03EA 95.75 CAD
CAAAB22866-03 CAAAB22866-06 CAAAB22866-14
1-Naphthyl phosphate disodium salt monohydrate 99%
1-Naphthyl phosphate disodium salt monohydrate

Purity: 99% CAS No.: 207569-06-0 Molecular Formula: C10H7Na2O4P.xH2O Formula Weight: 268.12(anhy) Storage Temperature: 2°C to 8°C Melting Point: 300° Beilstein Reference: 4895811 MDL No.: MFCD00150614

Formula: C₁₀H₇Na₂O₄P·1H₂O
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00150614
CAS Number: 207569-06-0
EINECS: 218-564-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR