(Phenylthio)acetic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

B22280-14 B22280-22
CAAAB22280-14EA 90.69 CAD
CAAAB22280-14 CAAAB22280-22
(Phenylthio)acetic acid 97%
(Phenylthio)acetic acid

Fieser: 6,463


Purity: 97% CAS No.: 103-04-8 Molecular Formula: C6H5SCH2CO2H Formula Weight: 168.22 Odor: Stench Melting Point: 61-66° Beilstein Reference: 2045072 MDL No.: MFCD00004355

Formula: C₈H₈O₂S
Melting Pt: 61…66 °C
Storage Temperature: Ambient
MDL Number: MFCD00004355
CAS Number: 103-04-8
EINECS: 203-073-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR