Rhodanine-N-acetic acid 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-(4-Oxo-2-thioxothiazolidin-3-yl)acetic acid, Rhodanine-3-acetic acid
B22244-06
B22244-14
B22244-22
CAAAB22244-06EA
47.24
CAD
CAAAB22244-06
CAAAB22244-14
CAAAB22244-22
Rhodanine-N-acetic acid 98%
Rhodanine-N-acetic acid
Purity: 98%
CAS No.: 5718-83-2
Molecular Formula: C5H5NO3S2
Formula Weight: 191.23
Melting Point: 145-148°
Beilstein Reference: 149513
Formula:
C₅H₅NO₃S₂ Melting Pt: 145…148 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005491 CAS Number: 5718-83-2 EINECS: 227-220-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...