Hydroxylamine 50% in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

B22202-AE B22202-AP
CAAAB22202-AEEA 77.26 CAD
CAAAB22202-AE CAAAB22202-AP
Hydroxylamine 50% in aqueous solution
Hydroxylamine

CAS No.: 7803-49-8 Molecular Formula: H3NO Formula Weight: 33.03 Flash Point: None Refractive Index: 1.3940 Odor: Weak Boiling Point: 107-114° Beilstein Reference: 1098214 Merck Reference: 14,4828 UN No.: 3266 MDL No.: MFCD00044522

Formula: H₃NO
MW: 33.03 g/mol
Boiling Pt: 107…114 °C
Melting Pt: 7 °C
Density: 1.120
Storage Temperature: Ambient
MDL Number: MFCD00044522
CAS Number: 7803-49-8
EINECS: 232-259-2
UN: 3266
ADR: 8,III
Merck Index: 14,4828

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR