Trifluoromethanesulfonamide 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22112-03
B22112-06
CAAAB22112-03EA
139.51
CAD
CAAAB22112-03
CAAAB22112-06
Trifluoromethanesulfonamide 96%
Trifluoromethanesulphonamide
Purity: 96%
CAS No.: 421-85-2
Molecular Formula: CF3SO2NH2
Formula Weight: 149.09
Melting Point: 116-119°
Beilstein Reference: 1812099
Formula:
CH₂F₃NO₂S Melting Pt: 116…119 °C Storage Temperature: Ambient |
MDL Number:
MFCD00068714 CAS Number: 421-85-2 EINECS: 431-270-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...