DL-Tropic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Hydroxy-2-phenylpropanoic acid, Tropic acid, 3-Hydroxy-2-phenylpropionic acid, (±)-Tropic acid, 2-Phenylhydracrylic acid
B22040-09
B22040-18
CAAAB22040-09EA
76.47
CAD
CAAAB22040-09
CAAAB22040-18
DL-Tropic acid 98%
DL-Tropic acid
Purity: 98%
CAS No.: 552-63-6
Molecular Formula: C9H10O3
Formula Weight: 166.18
Melting Point: 117-119°
Beilstein Reference: 2209199
Merck Reference: 14,9779
MDL No.: MFCD00004255
Formula:
C₉H₁₀O₃ MW: 166.18 g/mol Melting Pt: 117…119 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004255 CAS Number: 529-64-6 EINECS: 209-020-6 Merck Index: 14,9779 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...