3-(Methylthio)butanal 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22033-09
B22033-18
CAAAB22033-09EA
42.34
CAD
CAAAB22033-09
CAAAB22033-18
3-(Methylthio)butanal 95%
3-(Methylthio)butanal
Purity: 95%
CAS No.: 16630-52-7
Molecular Formula: CH3SCH(CH3)CH2CHO
Formula Weight: 118.20
Flash Point: 62°(144°F)
Boiling Point: 63-65°/11mm
MDL No.: MFCD00036622
Formula:
C₅H₁₀OS Boiling Pt: 63…65 °C/11 mm Flash Pt: 62 °C(144 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00036622 CAS Number: 16630-52-7 EINECS: 240-678-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...