(±)-Methyl-2,3-dichloropropionate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21993-14
B21993-22
B21993-36
CAAAB21993-14EA
82.95
CAD
CAAAB21993-14
CAAAB21993-22
CAAAB21993-36
(±)-Methyl-2,3-dichloropropionate 98%
(±)-Methyl 2,3-dichloropropionate
Fieser: 21,268
Purity: 98%
CAS No.: 3674-09-7
Molecular Formula: C4H6Cl2O2
Formula Weight: 157.00
Flash Point: 87°(188°F)
Refractive Index: 1.4530
Boiling Point: 91-92°/50mm
Beilstein Reference: 1701891
UN No.: 3265
MDL No.: MFCD00000944
Formula:
C₄H₆Cl₂O₂ Boiling Pt: 91…92 °C/50 mm Density: 1.326 Flash Pt: 87 °C(188 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000944 CAS Number: 3674-09-7 EINECS: 222-940-2 UN: 3265 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...