N-Acetylglycine(Ac-Gly-OH) 99%
Supplier: Thermo Scientific Chemicals
B21887-22
B21887-36
CAAAB21887-22EA
54.19
CAD
CAAAB21887-22
CAAAB21887-36
N-Acetylglycine(Ac-Gly-OH) 99%
N-Acetylglycine (Ac-Gly-OH)
Purity: 99%
CAS No.: 543-24-8
Molecular Formula: CH3CONHCH2CO2H
Formula Weight: 117.10
Melting Point: ca 210° dec.
Beilstein Reference: 774114
Merck Reference: 14,80
MDL No.: MFCD00004275
Formula:
C₄H₇NO₃ Melting Pt: ca 210 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00004275 CAS Number: 543-24-8 EINECS: 208-839-6 Merck Index: 14,80 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...