Rosolic acid (Aurin)

Supplier: Thermo Scientific Chemicals
Warning

B21737-14 B21737-22
CAAAB21737-14EA 134.3 CAD
CAAAB21737-14 CAAAB21737-22
Rosolic acid (Aurin)
Rosolic acid (Aurin)

CAS No.: 603-45-2 Molecular Formula: C19H14O3 Formula Weight: 290.32 Beilstein Reference: 2055205 Merck Reference: 14,881 MDL No.: MFCD00001624

Formula: C₁₉H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD00001624
CAS Number: 603-45-2
EINECS: 210-041-8
Merck Index: 14,881

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR