3-(2-Furyl)acrylic acid 99%
Supplier: Thermo Scientific Chemicals
B21653-30
B21653-14
B21653-22
CAAAAB21653-30EA
475.58
CAD
CAAAAB21653-30
CAAAB21653-14
CAAAB21653-22
3-(2-Furyl)acrylic acid 99%
3-(2-Furyl)acrylic acid
Purity: 99%
CAS No.: 539-47-9
Molecular Formula: C7H6O3
Formula Weight: 138.12
Melting Point: 140-144°
Boiling Point: 286°
Merck Reference: 14,4297
MDL No.: MFCD00003257
Formula:
C₇H₆O₃ Boiling Pt: 286 °C Melting Pt: 140…144 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003257 CAS Number: 539-47-9 EINECS: 208-718-8 Merck Index: 14,4297 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...