Ethyl-3-hydroxybutyrate 98+%
Supplier: Thermo Scientific Chemicals
Warning
|
B21521-14
B21521-22
CAAAB21521-14EA
66.52
CAD
CAAAB21521-14
CAAAB21521-22
Ethyl-3-hydroxybutyrate 98+%
Ethyl 3-hydroxybutyrate
Purity: 98+%
CAS No.: 5405-41-4
Molecular Formula: C6H12O3
Formula Weight: 132.16
Flash Point: 64°(148°F)
Refractive Index: 1.4210
Boiling Point: 169-171°
Beilstein Reference: 1446190
MDL No.: MFCD00004545
Formula:
C₆H₁₂O₃ Boiling Pt: 169…171 °C Density: 1.013 Flash Pt: 64 °C(148 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004545 CAS Number: 5405-41-4 EINECS: 226-456-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...