4,4,4-Trifluoro-2-butanone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B21514-03
B21514-06
CAAAB21514-03EA
98.43
CAD
CAAAB21514-03
CAAAB21514-06
4,4,4-Trifluoro-2-butanone 97%
4,4,4-Trifluoro-2-butanone
Purity: 97%
CAS No.: 2366-70-3
Molecular Formula: CF3CH2COCH3
Formula Weight: 126.08
Flash Point: 37°(99°F)
Boiling Point: 95-96°
UN No.: 1224
MDL No.: MFCD00077601
Formula:
C₄H₅F₃O Boiling Pt: 95…96 °C Density: 1.05 Flash Pt: 37 °C(99 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00077601 CAS Number: 2366-70-3 UN: 1224 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...