Selenourea 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B21440-03
B21440-06
CAAAB21440-03EA
140.46
CAD
CAAAB21440-03
CAAAB21440-06
Selenourea 99%
Selenourea
Purity: 99%
CAS No.: 630-10-4
Molecular Formula: (H2N)2C=Se
Formula Weight: 123.02
Storage Temperature: 2°C to 8°C
Melting Point: ca 210° dec.
UN No.: 3283
MDL No.: MFCD00008065
Formula:
CH₄N₂Se Melting Pt: ca 210 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00008065 CAS Number: 630-10-4 EINECS: 211-129-9 UN: 3283 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...