1,5-Dibromo-1,1,3,3,5,5-hexafluoropentane 97%

Supplier: Thermo Scientific Chemicals
Warning

B21405-04 B21405-09
CAAAB21405-04EA 82.32 CAD
CAAAB21405-04 CAAAB21405-09
1,5-Dibromo-1,1,3,3,5,5-hexafluoropentane 97%
1,5-Dibromo-1,1,3,3,5,5-hexafluoropentane

Purity: 97% CAS No.: 371-83-5 Molecular Formula: C5H4Br2F6 Formula Weight: 337.89 Refractive Index: 1.4632

Formula: C₅H₄Br₂F₆
Density: 2,003
Storage Temperature: Ambient
MDL Number: MFCD00236657
CAS Number: 371-83-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR