Fmoc-Pro-OH 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-proline, N-Fmoc-L-proline
B21081-06
B21081-14
CAAAB21081-06EA
90.06
CAD
CAAAB21081-06
CAAAB21081-14
Fmoc-Pro-OH 98%
Fmoc-Pro-OH
Purity: 98%
CAS No.: 71989-31-6
Molecular Formula: C20H19NO4
Formula Weight: 337.38
Storage Temperature: 2°C to 8°C
Melting Point: 113-117°
Beilstein Reference: 3596735
Optical Rotation: -40° (c=1 in ethyl acetate)
MDL No.: MFCD00037122
Formula:
C₂₀H₁₉NO₄ MW: 337.38 g/mol Melting Pt: 113…117 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037122 CAS Number: 71989-31-6 EINECS: 276-259-0 |
Specification Test Results
Optical rotation | -40° (c=1 in ethyl acetate) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...