4-Aminobutyraldehyde dimethyl acetal 98+%

Supplier: Thermo Scientific Chemicals
Danger

B21072-06 B21072-14
CAAAB21072-06EA 87.69 CAD
CAAAB21072-06 CAAAB21072-14
4-Aminobutyraldehyde dimethyl acetal 98+%
4-Aminobutyraldehyde dimethyl acetal

Purity: 98+% CAS No.: 19060-15-2 Molecular Formula: C6H15NO2 Formula Weight: 133.19 Flash Point: 59°(138°F) Refractive Index: 1.4300 Boiling Point: 85°/22mm UN No.: 2734 MDL No.: MFCD00189373

Formula: C₆H₁₅NO₂
Boiling Pt: 85 °C/22 mm
Flash Pt: 59 °C(138 °F)
Storage Temperature: Ambient
MDL Number: MFCD00189373
CAS Number: 19060-15-2
EINECS: 407-690-6
UN: 2734
ADR: "8",II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR