2-Methyl-5-phenyl-3-furoic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Methyl-5-phenylfuran-3-carboxylic acid
B20988-03
B20988-06
CAAAB20988-03EA
159.58
CAD
CAAAB20988-03
CAAAB20988-06
2-Methyl-5-phenyl-3-furoic acid 97%
2-Methyl-5-phenyl-3-furoic acid
Purity: 97%
CAS No.: 108124-17-0
Molecular Formula: C12H10O3
Formula Weight: 202.21
Melting Point: 181-184°
MDL No.: MFCD00221070
Formula:
C₁₂H₁₀O₃ MW: 202.21 g/mol Melting Pt: 181…184 °C Storage Temperature: Ambient |
MDL Number:
MFCD00221070 CAS Number: 108124-17-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...