1,2,3-Thiadiazole-4-carboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B20842-03
B20842-06
CAAAAB20842-MDEA
136.56
CAD
CAAAAB20842-MD
CAAAB20842-03
CAAAB20842-06
1,2,3-Thiadiazole-4-carboxylic acid 97%
1,2,3-Thiadiazole-4-carboxylic acid
Purity: 97%
CAS No.: 4100-13-4
Molecular Formula: C3H2N2O2S
Formula Weight: 130.12
Melting Point: 227-228°
Beilstein Reference: 113707
Formula:
C₃H₂N₂O₂S MW: 130.13 Melting Pt: 227…228 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052099 CAS Number: 4100-13-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...