Aluminium trifluoromethanesulfonate 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Aluminum tris(trifluoromethanesulfonate), Aluminum trifluoromethanesulphonate, Aluminum trifluoromethanesulfonate, Aluminum triflate, Aluminum tris(trifluoromethanesulphonate)
B20785-06
B20785-14
CAAAAB20785-06EA
96.85
CAD
CAAAAB20785-06
CAAAAB20785-14
Aluminium trifluoromethanesulfonate 99%
Aluminium trifluoromethanesulphonate
Purity: 99%
CAS No.: 74974-61-1
Molecular Formula: C3AlF9O9S3
Formula Weight: 474.20
Melting Point: ca 300°
UN No.: 3261
MDL No.: MFCD00143596
Formula:
C₃AlF₉O₉S₃ Melting Pt: ca 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00143596 CAS Number: 74974-61-1 UN: 3261 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...