Diethylene glycol monohexyl ether 97%

Supplier: Thermo Scientific Chemicals
Danger

B20774-0D B20774-0F B20774-AP
CAAAB20774-0DEA 804.22 CAD
CAAAB20774-0D CAAAB20774-0F CAAAB20774-AP
Diethylene glycol monohexyl ether 97%
Diethylene glycol monohexyl ether

Purity: 97% CAS No.: 112-59-4 Molecular Formula: C10H22O3 Formula Weight: 190.28 Flash Point: 123°(253°F) Refractive Index: 1.4370 Melting Point: -40° Boiling Point: 259-261° Beilstein Reference: 1743959 MDL No.: MFCD00010703

Formula: C₁₀H₂₂O₃
Boiling Pt: 259…261 °C
Melting Pt: –40 °C
Density: 0.932
Flash Pt: 123 °C(253 °F)
Storage Temperature: Ambient
MDL Number: MFCD00010703
CAS Number: 112-59-4
EINECS: 203-988-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR