(-)-Carveol (mixture of cis- and trans-isomers) 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
p-Mentha-6,8-dien-2-ol, (5R)-5-Isopropenyl-2-methyl-2-cyclohexenol
B20636-09
B20636-18
B20636-30
CAAAB20636-09EA
85.48
CAD
CAAAB20636-09
CAAAB20636-18
CAAAB20636-30
(-)-Carveol (mixture of cis- and trans-isomers) 97%
(-)-Carveol (mixture of cis- and trans-isomers)
Purity: 97%
CAS No.: 99-48-9
Molecular Formula: C10H16O
Formula Weight: 152.24
Flash Point: 98°(209°F)
Refractive Index: 1.4960
Boiling Point: 227°
Beilstein Reference: 6269416
MDL No.: MFCD00869995
Formula:
C₁₀H₁₆O MW: 152.24 g/mol Boiling Pt: 227 °C Density: 0.96 Flash Pt: 98 °C(209 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00869995 CAS Number: 99-48-9 EINECS: 202-757-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...